This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LRFN1 SIRNA Assays ›
Gene Symbol:
LRFN1
Gene Name:
leucine rich repeat and fibronectin type III domain containing 1
Gene Aliases:
SALM2
Chromosome Location:
Chr.19: 39306817 - 39320867 on Build GRCh38
UniGene ID:
Hs.97860
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 272044

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LRFN1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020862.1 NP_065913.1 1 569
XM_005259102.4 XP_005259159.1 4 769
XM_017027033.1 XP_016882522.1 4 884
GenBank mRNA AB040917.1 - 1 358

Back To Top

More Information


Gene Ontology Categories:

Function(s)

chemical synaptic transmission
axonogenesis

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-kn8lg:80/100.66.75.109:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0