This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF469 SIRNA Assays ›
Gene Symbol:
ZNF469
Gene Name:
zinc finger protein 469
Gene Aliases:
BCS, BCS1
Chromosome Location:
Chr.16: 88424805 - 88440757 on Build GRCh38
UniGene ID:
Hs.54925
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 272179

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF469
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127464.2 NP_001120936.2 2 8465
XM_017023784.1 XP_016879273.1 2 9699
XM_017023785.1 XP_016879274.1 2 8742
GenBank mRNA AB058761.2 - 1 4099

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-xql92:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline