This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SIRPG SIRNA Assays ›
Gene Symbol:
SIRPG
Gene Name:
signal regulatory protein gamma
Gene Aliases:
CD172g, SIRP-B2, SIRPB2, SIRPgamma, bA77C3.1
Chromosome Location:
Chr.20: 1629152 - 1686516 on Build GRCh38
UniGene ID:
Hs.590883
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 27234

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SIRPG
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039508.1 NP_001034597.1 5 970
NM_018556.3 NP_061026.2 6 1303
NM_080816.2 NP_543006.2 4 652
XM_005260749.3 XP_005260806.1 5 1021
XM_011529286.2 XP_011527588.1 6 1350
GenBank mRNA AB042624.1 - 6 1319
AK124224.1 - 1 3683
AK292745.1 - 6 1303
BC020629.1 - 4 629
BC064532.1 - 6 1264 5588038
DB046293.1 - 3 336

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
cell-cell signaling
positive regulation of cell proliferation
negative regulation of cell proliferation
positive regulation of cell-cell adhesion
intracellular signal transduction
positive regulation of T cell activation
leukocyte migration
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-n7slw:80/100.66.78.89:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0