This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZFP92 SIRNA Assays ›
Gene Symbol:
ZFP92
Gene Name:
ZFP92 zinc finger protein
Gene Aliases:
ZNF897
Chromosome Location:
Chr.X: 153411461 - 153426481 on Build GRCh38
UniGene ID:
Hs.534554
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 272958

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZFP92
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136273.1 NP_001129745.1 3 185
XM_005274652.3 XP_005274709.1 5 495
XM_011531115.2 XP_011529417.1 4 446
XM_017029281.1 XP_016884770.1 6 844

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0