This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RNF169 SIRNA Assays ›
Gene Symbol:
RNF169
Gene Name:
ring finger protein 169
Gene Aliases:
-
Chromosome Location:
Chr.11: 74748856 - 74842413 on Build GRCh38
UniGene ID:
Hs.370145
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 273449

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RNF169
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098638.1 NP_001092108.1 6 2151
XM_011544889.2 XP_011543191.1 6 2226
GenBank mRNA AB082522.1 - 6 2151
BX640750.1 - 1 1055

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cellular response to DNA damage stimulus
protein ubiquitination
negative regulation of double-strand break repair
zinc ion binding
ligase activity
nucleosome binding
K63-linked polyubiquitin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-jq955:80/100.66.72.86:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0