This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SUSD5 SIRNA Assays ›
Gene Symbol:
SUSD5
Gene Name:
sushi domain containing 5
Gene Aliases:
-
Chromosome Location:
Chr.3: 33150041 - 33219215 on Build GRCh38
UniGene ID:
Hs.196647
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 273517

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 286.00
Your Price
Online offer: 277.65
277.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SUSD5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015551.1 NP_056366.1 5 3041
XM_005265034.3 XP_005265091.1 4 2589
XM_017006137.1 XP_016861626.1 5 2991
GenBank mRNA AB011099.1 - 5 3041

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
Notch signaling pathway
hyaluronic acid binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline