siRNA Details
| Gene Symbol: LINC00486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027098.1 | 5 | 1164 | |||
| NR_027100.1 | 4 | 901 | ||||
| GenBank mRNA | AK095182.1 | - | 5 | 1164 | ||
| BC121144.1 | - | 4 | 901 | |||
| BC121145.1 | - | 4 | 904 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00486| Gene Symbol: LINC00486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027098.1 | 5 | 1164 | |||
| NR_027100.1 | 4 | 901 | ||||
| GenBank mRNA | AK095182.1 | - | 5 | 1164 | ||
| BC121144.1 | - | 4 | 901 | |||
| BC121145.1 | - | 4 | 904 | |||