This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SPRED3 SIRNA Assays ›
Gene Symbol:
SPRED3
Gene Name:
sprouty related EVH1 domain containing 3
Gene Aliases:
Eve-3, spred-3
Chromosome Location:
Chr.19: 38390061 - 38399884 on Build GRCh38
UniGene ID:
Hs.196011
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 275855

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SPRED3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042522.2 NP_001035987.1 2 369
NR_073032.1 2 184
XM_006723219.3 XP_006723282.1 3 360
XM_011526978.2 XP_011525280.1 2 415
GenBank mRNA BC151136.1 - 2 369 9057036
BF983829.1 - 2 186
DQ323927.1 - 2 270
DQ323928.1 - 2 270

Back To Top

More Information


Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s)

multicellular organism development
regulation of signal transduction

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9hvhj:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline