siRNA Details
Gene Symbol: TMEM191C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207052.1 | NP_001193981.1 | 9 | 1288 | ||
GenBank mRNA | AK094309.1 | - | 7 | 881 | ||
AK094490.1 | - | 7 | 1493 | |||
AK308704.1 | - | 6 | 469 | |||
AL041322.1 | - | 5 | 610 |
Gene Symbol: TMEM191B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242313.1 | NP_001229242.1 | 9 | 1315 | ||
XM_011546160.2 | XP_011544462.1 | 10 | 1321 | |||
GenBank mRNA | AK093171.1 | - | 5 | 1575 |
