siRNA Details
| Gene Symbol: LOC101928307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB073651.1 | - | 1 | 172 | ||
| AL833240.1 | - | 1 | 333 | |||
| DA296736.1 | - | 2 | 179 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928307| Gene Symbol: LOC101928307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB073651.1 | - | 1 | 172 | ||
| AL833240.1 | - | 1 | 333 | |||
| DA296736.1 | - | 2 | 179 | |||