This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other KRTAP2-2 SIRNA Assays ›
Gene Symbol:
KRTAP2-2 KRTAP2-4
Gene Name:
keratin associated protein 2-2
keratin associated protein 2-4
Gene Aliases:
KAP2.2
KAP2.1B, KAP2.4, KRTAP2.4
Chromosome Location:
Chr.17: 41054498 - 41055211 on Build GRCh38
Chromosome Location:
Chr.17: 41065116 - 41065879 on Build GRCh38
UniGene ID:
Hs.650131
Hs.560502
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID 278969

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KRTAP2-2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033032.2 NP_149021.2 1 603
GenBank mRNA AJ302536.1 - 490
Gene Symbol: KRTAP2-4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AJ406929.1 - 424

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline