siRNA Details
| Gene Symbol: LINC01620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132342.1 | 2 | 211 | |||
| NR_132343.1 | 2 | 211 | ||||
| GenBank mRNA | BC030259.1 | - | 2 | 212 | ||
| BX114953.1 | - | 2 | 195 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01620| Gene Symbol: LINC01620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132342.1 | 2 | 211 | |||
| NR_132343.1 | 2 | 211 | ||||
| GenBank mRNA | BC030259.1 | - | 2 | 212 | ||
| BX114953.1 | - | 2 | 195 | |||