This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CEP170B SIRNA Assays ›
Gene Symbol:
CEP170B
Gene Name:
centrosomal protein 170B
Gene Aliases:
CEP170R, FAM68C, KIAA0284
Chromosome Location:
Chr.14: 104865313 - 104896770 on Build GRCh38
UniGene ID:
Hs.533721
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 279677

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CEP170B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001112726.2 NP_001106197.1 15, 16 4409
NM_015005.2 NP_055820.2 14, 15 4382
XM_005267550.3 XP_005267607.1 15, 16 4496
XM_011536665.2 XP_011534967.1 16, 17 4649
XM_011536666.2 XP_011534968.1 15, 16 5012
XM_011536667.2 XP_011534969.1 8, 9 2688
XM_017021227.1 XP_016876716.1 14, 15 5261
XM_017021228.1 XP_016876717.1 14, 15 5156
GenBank mRNA AB006622.3 - 15, 16 4409
AK091980.1 - 3, 4 2018
AK300834.1 - 1, 2 170
BC047913.1 - 1, 2 77
BC112928.1 - 14, 15 4413 6141521

Back To Top

More Information


Panther Classification:

Molecular Function -

scaffold/adaptor protein

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline