This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other C12ORF74 SIRNA Assays ›
Gene Symbol:
C12orf74
Gene Name:
chromosome 12 open reading frame 74
Gene Aliases:
-
Chromosome Location:
Chr.12: 92702843 - 92708549 on Build GRCh38
UniGene ID:
Hs.326303
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 280045

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C12orf74
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037671.3 NP_001032760.1 2 867
NM_001178097.2 NP_001171568.1 2 867
GenBank mRNA BC043363.1 - 2 682
BC043364.1 - 2 682
BC105728.1 - 2 647 40025557
BC107813.1 - 2 646 40025555

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0