siRNA Details
| Gene Symbol: Usp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025424.1 | NP_001020595.1 | 15 | 1538 | ||
| GenBank mRNA | BC129075.1 | - | 14 | 1753 | 7383067 | |
| DQ076481.1 | - | 15 | 1538 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp3| Gene Symbol: Usp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025424.1 | NP_001020595.1 | 15 | 1538 | ||
| GenBank mRNA | BC129075.1 | - | 14 | 1753 | 7383067 | |
| DQ076481.1 | - | 15 | 1538 | |||