This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SP110 SIRNA Assays ›
Gene Symbol:
Sp110
Gene Name:
SP110 nuclear body protein
Gene Aliases:
-
Chromosome Location:
Chr.9: 92593909 - 92618808 on Build Rnor_6.0
UniGene ID:
Rn.6459
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 281686

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sp110
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034137.1 NP_001029309.1 12, 13 1494
XM_006245370.2 XP_006245432.1 11, 12 1375
XM_008767234.1 XP_008765456.1 13, 14 1558
GenBank mRNA BC105751.1 - 12, 13 1494 5599224

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
response to bacterium
positive regulation of apoptotic process
innate immune response
DNA binding
chromatin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline