This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FUZ SIRNA Assays ›
Gene Symbol:
Fuz
Gene Name:
fuzzy planar cell polarity protein
Gene Aliases:
RGD1310608
Chromosome Location:
Chr.1: 100891826 - 100896811 on Build Rnor_6.0
UniGene ID:
Rn.136607
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 281836

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fuz
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037646.1 NP_001032735.1 2, 3 381
XM_006229059.2 XP_006229121.1 2, 3 420
XM_006229060.2 XP_006229122.1 2, 3 420
XM_008759373.1 XP_008757595.1 2, 3 421
XM_008759374.1 XP_008757596.1 2, 3 421
GenBank mRNA BC107809.1 - 2, 3 381 7453736
FQ228777.1 - 2, 3 366

Back To Top

More Information


Gene Ontology Categories:

Function(s)

establishment of planar polarity
neural tube closure
hair follicle development
negative regulation of cell proliferation
regulation of smoothened signaling pathway
embryonic body morphogenesis
protein transport
spinal cord dorsal/ventral patterning
negative regulation of cell migration
nonmotile primary cilium assembly
cilium assembly
positive regulation of cilium assembly
embryonic skeletal system morphogenesis
negative regulation of canonical Wnt signaling pathway
negative regulation of neural crest formation
negative regulation of fibroblast growth factor receptor signaling pathway involved in neural plate anterior/posterior pattern formation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-kbrl8:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline