This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZC3H18 SIRNA Assays ›
Gene Symbol:
Zc3h18
Gene Name:
zinc finger CCCH-type containing 18
Gene Aliases:
1190001B23Rik, 5830416A07Rik, Nhn1
Chromosome Location:
Chr.8: 122376614 - 122417360 on Build GRCm38
UniGene ID:
Mm.241489
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 282716

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zc3h18
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029993.1 NP_001025164.1 7 1253
NM_001029994.1 NP_001025165.1 6 1181
GenBank mRNA AK031413.1 - 7 1202
AK131189.1 - 4 325
AK159881.1 - 7 1189
AK165989.1 - 5 532
BC030495.1 - 6 1161 5400494
BC094330.1 - 4 334

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_201416, XM_008772603, XM_006255758, XM_006255760, XM_006255761, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline