siRNA Details
| Gene Symbol: Olfr1318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011802.2 | NP_001011802.1 | 1 | 948 | ||
| GenBank mRNA | BC147163.1 | - | 1 | 948 | 9056186 | |
| BC147164.1 | - | 1 | 948 | 9056168 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1318| Gene Symbol: Olfr1318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011802.2 | NP_001011802.1 | 1 | 948 | ||
| GenBank mRNA | BC147163.1 | - | 1 | 948 | 9056186 | |
| BC147164.1 | - | 1 | 948 | 9056168 | ||