siRNA Details
| Gene Symbol: Lonrf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028894.1 | NP_083170.1 | 6, 7 | 1679 | ||
| GenBank mRNA | AK016522.1 | - | 6, 7 | 1679 | ||
| AK167341.1 | - | 6, 7 | 1737 | |||
| BC120688.1 | - | 6, 7 | 1581 | 40129611 | ||
| BC120690.1 | - | 6, 7 | 1581 | 40129613 | ||

Lonrf3
Genomic Map