This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SIX5 SIRNA Assays ›
Gene Symbol:
Six5
Gene Name:
sine oculis-related homeobox 5
Gene Aliases:
Dmahp, MDMAHP, TrexBF
Chromosome Location:
Chr.7: 19094544 - 19098549 on Build GRCm38
UniGene ID:
Mm.3410
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 284024

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Six5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011383.1 NP_035513.1 2 1214
GenBank mRNA D83146.1 - 2 965

Back To Top

More Information


Panther Classification:

Molecular Function -

homeodomain transcription factor

Gene Ontology Categories:

Function(s) Process(es)

lens development in camera-type eye
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
spermatid development
negative regulation of cell proliferation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of skeletal muscle satellite cell proliferation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline