siRNA Details
| Gene Symbol: Wdr93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037927.1 | NP_001033016.1 | 3 | 532 | ||
| GenBank mRNA | AB205021.1 | - | 3 | 532 | ||
| BC150920.1 | - | 2 | 502 | 9087831 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr93| Gene Symbol: Wdr93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037927.1 | NP_001033016.1 | 3 | 532 | ||
| GenBank mRNA | AB205021.1 | - | 3 | 532 | ||
| BC150920.1 | - | 2 | 502 | 9087831 | ||