This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CSAG3 SIRNA Assays ›
Gene Symbol:
CSAG3 CSAG2
Gene Name:
CSAG family member 3
CSAG family member 2
Gene Aliases:
CSAG3A, CT24.2
CSAG3B, CT24.2, TRAG-3, TRAG3
Chromosome Location:
Chr.X: 152753921 - 152760222 on Build GRCh38
Chromosome Location:
Chr.X: 152708261 - 152713861 on Build GRCh38
UniGene ID:
Hs.721609
Hs.735281
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID 284722

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CSAG3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001129826.2 NP_001123298.1 1 186
NM_001129828.2 NP_001123300.1 1 186
GenBank mRNA AF080246.1 - 1 186
AF136715.1 - 1 186
BC119734.1 - 1 159 40119148
BC119735.1 - 1 158 40119150
Gene Symbol: CSAG2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006724857.3 XP_006724920.1 1 312
XM_017030023.1 XP_016885512.1 1 312

Back To Top

More Information


Gene Ontology Categories:

Function(s)

response to drug

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-2x92s:80/100.66.79.168:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0