This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CFAP206 SIRNA Assays ›
Gene Symbol:
CFAP206
Gene Name:
cilia and flagella associated protein 206
Gene Aliases:
C6orf165, dJ382I10.1
Chromosome Location:
Chr.6: 87407972 - 87464473 on Build GRCh38
UniGene ID:
Hs.82921
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 284944

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CFAP206
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031743.2 NP_001026913.1 2 198
GenBank mRNA AK093830.1 - 2 157
BC035083.2 - 2 167 5260875
DB021694.1 - 2 194

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-w8hc8:80/100.66.77.245:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0