siRNA Details
| Gene Symbol: Olr1470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000723.1 | NP_001000723.1 | 1 | 705 | ||
| GenBank mRNA | AF091569.1 | - | 1 | 516 | ||
| M64388.1 | - | 1 | 705 | |||
| Gene Symbol: LOC100909493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750851.3 | XP_003750899.1 | 1 | 705 | ||

Olr1470
Genomic Map