This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DEFA10 SIRNA Assays ›
Gene Symbol:
Defa10
Gene Name:
defensin alpha 10
Gene Aliases:
-
Chromosome Location:
Chr.16: 75498213 - 75500336 on Build Rnor_6.0
UniGene ID:
Rn.215840
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 286697

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Defa10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033074.1 NP_001028246.1 2 249
XM_008771364.1 XP_008769586.1 3 450
XM_008771365.1 XP_008769587.1 3 447
GenBank mRNA AY623754.1 - 2 249

Back To Top

More Information


Panther Classification:

Molecular Function -

antimicrobial response protein

Gene Ontology Categories:

Function(s)

defense response to bacterium

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline