siRNA Details
| Gene Symbol: Bcl7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009746.2 | NP_033876.1 | 5 | 748 | ||
| GenBank mRNA | AK049064.1 | - | 4 | 626 | ||
| AK131935.1 | - | 5 | 637 | |||
| BC002252.1 | - | 2 | 221 | |||
| BC005673.1 | - | 5 | 748 | 3153237 | ||
| BC058530.1 | - | 5 | 619 | 6818556 | ||
| Y11905.1 | - | 6 | 566 | |||

Bcl7c
Genomic Map