siRNA Details
| Gene Symbol: Ccdc172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029372.2 | NP_083648.1 | 3, 4 | 284 | ||
| GenBank mRNA | AK005864.1 | - | 3, 4 | 256 | ||
| BC049707.1 | - | 3, 4 | 264 | 6740418 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc172| Gene Symbol: Ccdc172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029372.2 | NP_083648.1 | 3, 4 | 284 | ||
| GenBank mRNA | AK005864.1 | - | 3, 4 | 256 | ||
| BC049707.1 | - | 3, 4 | 264 | 6740418 | ||