This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LRRN1 SIRNA Assays ›
Gene Symbol:
LRRN1
Gene Name:
leucine rich repeat neuronal 1
Gene Aliases:
FIGLER3, NLRR-1
Chromosome Location:
Chr.3: 3799431 - 3849834 on Build GRCh38
UniGene ID:
Hs.163244
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 28799

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LRRN1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324188.1 NP_001311117.1 3 2178
NM_001324189.1 NP_001311118.1 3 2101
NM_020873.6 NP_065924.3 2 1896
GenBank mRNA AB040930.1 - 2 4395
AB074161.1 - 1 1129
AK023273.1 - 1 283
AK291717.1 - 2 1480
AY358312.1 - 1 1243
BC034947.2 - 2 1459 4826337

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s)

axonogenesis
positive regulation of synapse assembly

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline