siRNA Details
| Gene Symbol: TNP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005425.4 | NP_005416.1 | 2 | 587 | ||
| GenBank mRNA | X63759.1 | - | 1 | 1663 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNP2| Gene Symbol: TNP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005425.4 | NP_005416.1 | 2 | 587 | ||
| GenBank mRNA | X63759.1 | - | 1 | 1663 | ||