siRNA Details
| Gene Symbol: ZNF138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160183.2 | NP_001153655.1 | 5 | 1435 | ||
| NM_001271637.1 | NP_001258566.1 | 4 | 1302 | |||
| NM_001271638.1 | NP_001258567.1 | 4 | 1309 | |||
| NM_001271639.1 | NP_001258568.1 | 4 | 1298 | |||
| NM_001271640.1 | NP_001258569.1 | 4 | 1357 | |||
| NM_001271649.1 | NP_001258578.1 | 2 | 1093 | |||
| NM_006524.3 | NP_006515.2 | 3 | 1220 | |||
| NR_073391.1 | 5 | 1402 | ||||
| GenBank mRNA | AK055644.1 | - | 4 | 1262 | ||
| BC131762.1 | - | 3 | 1184 | 40121140 | ||
| BC141847.1 | - | 4 | 1269 | |||

ZNF138
Genomic Map