This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF510 SIRNA Assays ›
Gene Symbol:
ZNF510
Gene Name:
zinc finger protein 510
Gene Aliases:
-
Chromosome Location:
Chr.9: 96754549 - 96778549 on Build GRCh38
UniGene ID:
Hs.633105
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 289719

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF510
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001314059.1 NP_001300988.1 5 993
NM_001314060.1 NP_001300989.1 4 545
NM_014930.2 NP_055745.1 5 604
XM_005251808.2 XP_005251865.1 5 594
XM_011518393.2 XP_011516695.1 5 1049
XM_017014483.1 XP_016869972.1 6 656
GenBank mRNA AB023189.1 - 4 521
AK292600.1 - 5 574
BC036676.1 - 5 991 4792684
BC058022.1 - 5 568
BC068587.1 - 5 987 4374918
DA496439.1 - 4 483

Back To Top

More Information


Panther Classification:

Molecular Function -

DNA-binding transcription factor C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline