This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
See other IP6K2 SIRNA Assays ›
Gene Symbol:
IP6K2
Gene Name:
inositol hexakisphosphate kinase 2
Gene Aliases:
IHPK2, PIUS
Chromosome Location:
Chr.3: 48688003 - 48717278 on Build GRCh38
UniGene ID:
Hs.595983
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
289762
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IP6K2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005909.2
NP_001005909.1
2
317
NM_001005910.2
NP_001005910.1
2
320
NM_001005911.2
NP_001005911.1
2
317
NM_001146178.2
NP_001139650.1
2
320
NM_001146179.2
NP_001139651.1
2
317
NM_001190316.1
NP_001177245.1
3
533
NM_001190317.1
NP_001177246.1
3
456
NM_016291.3
NP_057375.2
2
320
NR_027437.2
2
320
NR_027438.2
2
317
XM_006713199.3
XP_006713262.1
3
424
XM_006713200.1
XP_006713263.1
3
394
XM_006713201.1
XP_006713264.1
3
471
XM_006713202.1
XP_006713265.1
3
473
XM_011533816.1
XP_011532118.1
3
454
XM_011533817.1
XP_011532119.1
2
257
XM_011533818.1
XP_011532120.1
2
260
XM_011533822.1
XP_011532124.1
2
257
XM_011533823.1
XP_011532125.1
2
260
XM_017006583.1
XP_016862072.1
2
316
XM_017006584.1
XP_016862073.1
2
291
XM_017006585.1
XP_016862074.1
3
406
XM_017006586.1
XP_016862075.1
3
447
XM_017006587.1
XP_016862076.1
3
447
XM_017006588.1
XP_016862077.1
3
473
XM_017006589.1
XP_016862078.1
3
422
XM_017006590.1
XP_016862079.1
3
1028
XM_017006591.1
XP_016862080.1
3
393
XM_017006592.1
XP_016862081.1
3
422
XM_017006593.1
XP_016862082.1
3
422
GenBank mRNA
AF177145.1
-
1
214
AK024045.1
-
2
257
AK097016.1
-
8
1175
AK290526.1
-
3
471
AK291501.1
-
3
448
AK304714.1
-
3
394
AK315204.1
-
2
258
AL137514.1
-
2
258
AY007091.1
-
2
243
BC001864.1
-
2
242
BC004469.2
-
2
247
BC019694.1
-
2
229
4941646
BC065533.1
-
2
246
6162048
BP262706.1
-
2
257
BX537544.1
-
3
472
CA488567.1
-
2
254
CD248129.1
-
2
283
CF139656.1
-
2
251
CN362780.1
-
2
232
DA083858.1
-
2
253
DB039955.1
-
2
317
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
phosphate ion transport
negative regulation of cell growth
positive regulation of apoptotic process
inositol phosphate metabolic process
phosphatidylinositol phosphorylation
type I interferon signaling pathway
inositol-1,3,4,5,6-pentakisphosphate kinase activity
inositol hexakisphosphate kinase activity
inositol heptakisphosphate kinase activity
inositol hexakisphosphate 5-kinase activity
protein binding
ATP binding
inositol-1,4,5-trisphosphate 3-kinase activity
inositol hexakisphosphate 1-kinase activity
inositol hexakisphosphate 3-kinase activity
inositol 5-diphosphate pentakisphosphate 5-kinase activity
inositol diphosphate tetrakisphosphate kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67f85b6989-7wnc4:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline