This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
See other IP6K2 SIRNA Assays ›
Gene Symbol:
IP6K2
Gene Name:
inositol hexakisphosphate kinase 2
Gene Aliases:
IHPK2, PIUS
Chromosome Location:
Chr.3: 48688003 - 48717278 on Build GRCh38
UniGene ID:
Hs.595983
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
289763
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IP6K2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005909.2
NP_001005909.1
2
267
NM_001005910.2
NP_001005910.1
2
270
NM_001005911.2
NP_001005911.1
2
267
NM_001146178.2
NP_001139650.1
2
270
NM_001146179.2
NP_001139651.1
2
267
NM_001190316.1
NP_001177245.1
3
483
NM_001190317.1
NP_001177246.1
3
406
NM_016291.3
NP_057375.2
2
270
NR_027437.2
2
270
NR_027438.2
2
267
XM_006713199.3
XP_006713262.1
3
374
XM_006713200.1
XP_006713263.1
3
344
XM_006713201.1
XP_006713264.1
3
421
XM_006713202.1
XP_006713265.1
3
423
XM_011533816.1
XP_011532118.1
3
404
XM_011533817.1
XP_011532119.1
2
207
XM_011533818.1
XP_011532120.1
2
210
XM_011533822.1
XP_011532124.1
2
207
XM_011533823.1
XP_011532125.1
2
210
XM_017006583.1
XP_016862072.1
2
266
XM_017006584.1
XP_016862073.1
2
241
XM_017006585.1
XP_016862074.1
3
356
XM_017006586.1
XP_016862075.1
3
397
XM_017006587.1
XP_016862076.1
3
397
XM_017006588.1
XP_016862077.1
3
423
XM_017006589.1
XP_016862078.1
3
372
XM_017006590.1
XP_016862079.1
3
978
XM_017006591.1
XP_016862080.1
3
343
XM_017006592.1
XP_016862081.1
3
372
XM_017006593.1
XP_016862082.1
3
372
GenBank mRNA
AF177145.1
-
1
164
AK024045.1
-
2
207
AK097016.1
-
8
1125
AK290526.1
-
3
421
AK291501.1
-
3
398
AK304714.1
-
3
344
AK315204.1
-
2
208
AL137514.1
-
2
208
AY007091.1
-
2
193
BC001864.1
-
2
192
BC004469.2
-
2
197
BC019694.1
-
2
179
4941646
BC065533.1
-
2
196
6162048
BP262706.1
-
2
207
BX537544.1
-
3
422
CA488567.1
-
2
204
CB144374.1
-
2
265
CD248129.1
-
2
233
CF139656.1
-
2
201
CN362780.1
-
2
182
DA083858.1
-
2
203
DB039955.1
-
2
267
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
phosphate ion transport
negative regulation of cell growth
positive regulation of apoptotic process
inositol phosphate metabolic process
phosphatidylinositol phosphorylation
type I interferon signaling pathway
inositol-1,3,4,5,6-pentakisphosphate kinase activity
inositol hexakisphosphate kinase activity
inositol heptakisphosphate kinase activity
inositol hexakisphosphate 5-kinase activity
protein binding
ATP binding
inositol-1,4,5-trisphosphate 3-kinase activity
inositol hexakisphosphate 1-kinase activity
inositol hexakisphosphate 3-kinase activity
inositol 5-diphosphate pentakisphosphate 5-kinase activity
inositol diphosphate tetrakisphosphate kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67f85b6989-xxb7z:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline