This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MAP3K19 SIRNA Assays ›
Gene Symbol:
MAP3K19
Gene Name:
mitogen-activated protein kinase kinase kinase 19
Gene Aliases:
RCK, YSK4
Chromosome Location:
Chr.2: 134963875 - 135047468 on Build GRCh38
UniGene ID:
Hs.659395
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 289951

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MAP3K19
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018046.2 NP_001018056.1 4 329
NM_001018047.2 NP_001018057.1 4 329
NM_001282883.1 NP_001269812.1 4 329
NM_001321177.1 NP_001308106.1 4 469
NM_025052.4 NP_079328.3 4 329
XM_011511891.2 XP_011510193.1 4 469
XM_011511892.1 XP_011510194.1 7 1037
XM_011511894.2 XP_011510196.1 4 444
XM_011511896.2 XP_011510198.1 4 310
XM_011511897.2 XP_011510199.1 4 390
XM_017005003.1 XP_016860492.1 7 1004
XM_017005004.1 XP_016860493.1 6 889
XM_017005005.1 XP_016860494.1 5 690
XM_017005006.1 XP_016860495.1 4 469
XM_017005007.1 XP_016860496.1 4 469
GenBank mRNA AK122935.1 - 4 469
AK309454.1 - 6 889
AY574901.1 - 4 329
AY574903.1 - 4 329
AY574904.1 - 4 329
AY574905.1 - 4 329
BC137276.1 - 4 328 9021273
BC137277.1 - 4 328 9021274
BC144555.1 - 4 328

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal transduction by protein phosphorylation
actin cytoskeleton organization
receptor signaling protein serine/threonine kinase activity
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline