siRNA Details
| Gene Symbol: ELSPBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022142.4 | NP_071425.3 | 3 | 267 | ||
| XM_017027130.1 | XP_016882619.1 | 3 | 366 | |||
| GenBank mRNA | AF279147.1 | - | 3 | 267 | ||
| AJ278478.1 | - | 2 | 108 | |||
| BC015598.2 | - | 3 | 231 | 4649702 | ||
| GQ149084.1 | - | 3 | 231 | |||

ELSPBP1
Genomic Map