This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CRLF2 SIRNA Assays ›
Gene Symbol:
CRLF2
Gene Name:
cytokine receptor-like factor 2
Gene Aliases:
CRL2, CRLF2Y, TSLPR
Chromosome Location:
Chr.X: 1190437 - 1212762 on Build GRCh38
UniGene ID:
Hs.287729
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 29863

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CRLF2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022148.3 NP_071431.2 2 182
NR_110830.1 1, 2 179
XM_011546181.2 XP_011544483.1 1, 2 217
XM_011546182.2 XP_011544484.1 2 221
GenBank mRNA AB052639.1 - 2 93
AF142570.1 - 2 109
AF338733.1 - 2 93
AK026800.1 - 1, 2 181
AK223303.1 - 2 209
AK225583.1 - 1, 2 181

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4m7g7:80/100.66.75.155:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline