This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other NAA16 SIRNA Assays ›
Gene Symbol:
NAA16
Gene Name:
N(alpha)-acetyltransferase 16, NatA auxiliary subunit
Gene Aliases:
NARG1L
Chromosome Location:
Chr.13: 41311205 - 41377031 on Build GRCh38
UniGene ID:
Hs.512914
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 31002

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NAA16
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024561.4 NP_078837.3 15 2147
XM_005266523.2 XP_005266580.1 8 999
XM_006719866.2 XP_006719929.1 13 1943
XM_011535227.2 XP_011533529.1 15 2123
XM_017020744.1 XP_016876233.1 11 1466
GenBank mRNA AK025707.1 - 2 185
AK291248.1 - 15 2061
AL833341.1 - 9 1124
BC017099.1 - 5 733
BC028112.1 - 5 734
BC101719.1 - 15 1919 8069225
BC101721.1 - 15 1919 8069227
BX640665.1 - 16 2184

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

acetyltransferase

Gene Ontology Categories:

Function(s) Process(es)

N-terminal protein amino acid acetylation
N-terminal peptidyl-methionine acetylation
negative regulation of apoptotic process
positive regulation of transcription, DNA-templated
protein stabilization
peptide alpha-N-acetyltransferase activity
protein binding
acetyltransferase activity
ribosome binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline