This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DHRS12 SIRNA Assays ›
Gene Symbol:
DHRS12
Gene Name:
dehydrogenase/reductase 12
Gene Aliases:
SDR40C1
Chromosome Location:
Chr.13: 51754856 - 51804206 on Build GRCh38
UniGene ID:
Hs.266728
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 31410

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DHRS12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031719.2 NP_001026889.1 4 357
NM_001270424.1 NP_001257353.1 5 440
NM_024705.2 NP_078981.1 4 357
XM_005266527.3 XP_005266584.1 4 363
XM_005266528.4 XP_005266585.1 5 484
XM_011535235.2 XP_011533537.1 5 484
XM_011535236.2 XP_011533538.1 5 484
XM_011535237.1 XP_011533539.1 4 363
XM_011535238.1 XP_011533540.1 4 363
XM_017020749.1 XP_016876238.1 5 484
XM_017020750.1 XP_016876239.1 5 484
XM_017020751.1 XP_016876240.1 4 363
XM_017020752.1 XP_016876241.1 4 363
XM_017020753.1 XP_016876242.1 4 363
XM_017020754.1 XP_016876243.1 4 359
GenBank mRNA AK023701.1 - 4 352
BC009825.2 - 4 293 3942115
BC026024.1 - 5 373

Back To Top

More Information


Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
oxidoreductase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-584499df9d-gjjbx:80/100.66.78.233:80.
git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0