This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DPY19L2P2 SIRNA Assays ›
Gene Symbol:
DPY19L2P2 AP5Z1 CES3 LINC01447 LOC100130657 LOC101928336 LOC101929333
Gene Name:
DPY19L2 pseudogene 2
adaptor related protein complex 5 zeta 1 subunit
carboxylesterase 3
long intergenic non-protein coding RNA 1447
uncharacterized LOC100130657
uncharacterized LOC101928336
uncharacterized LOC101929333
Gene Aliases:
-
KIAA0415, SPG48, zeta
ES31
-
-
-
-
Chromosome Location:
Chr.7: 103175013 - 103280466 on Build GRCh38
Chromosome Location:
Chr.7: 4775617 - 4794395 on Build GRCh38
Chromosome Location:
Chr.16: 66961229 - 66975152 on Build GRCh38
Chromosome Location:
Chr.7: 47621939 - 47629893 on Build GRCh38
Chromosome Location:
-
Chromosome Location:
Chr.X: 119291529 - 119335610 on Build GRCh38
Chromosome Location:
Chr.15: 74478068 - 74490286 on Build GRCh38
UniGene ID:
Hs.732579
Hs.558440
Hs.268700
Hs.563487
Hs.679182
-
-
Species:
Human
Species Specific ID (Flybase ID):
-
-
-
-
-
-
-
siRNA ID 32060

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DPY19L2P2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003561.2 5 1401
NR_027768.1 3 1696
GenBank mRNA AK097711.1 - 3 1624
AK131353.1 - 5 1394
AL834175.1 - 3 1449
BC023575.2 - 4 1798
BC068601.1 - 3 1507
BC150309.1 - 3 1157
BX648212.1 - 4 1678
Gene Symbol: AP5Z1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014855.2 NP_055670.1 17 5169
GenBank mRNA AB007875.1 - 17 5393
Gene Symbol: CES3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001185176.1 NP_001172105.1 6 2813
NM_001185177.1 NP_001172106.1 13 3520
NM_024922.5 NP_079198.2 13 3529
XM_011522975.2 XP_011521277.1 13 3601
GenBank mRNA AK025158.1 - 3 2111
AK025171.1 - 1 1392
AK025389.1 - 6 2812
AY358609.1 - 13 3455
Gene Symbol: LINC01447
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_108090.1 4 1901
GenBank mRNA AK056440.1 - 4 1901
Gene Symbol: LOC100130657
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK126078.1 - 1 1224
Gene Symbol: LOC101928336
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_110396.1 4 1449
GenBank mRNA BC041420.1 - 4 1449
Gene Symbol: LOC101929333
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK127909.1 - 2 1601

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

esterase

Gene Ontology Categories:

Function(s) Process(es)

spermatid development
protein C-linked glycosylation via 2'-alpha-mannosyl-L-tryptophan
double-strand break repair via homologous recombination
protein transport
endosomal transport
mannosyltransferase activity
protein binding
carboxylic ester hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9vh6g:80/100.66.76.55:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline