siRNA Details
| Gene Symbol: GOLGA2P10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK026448.1 | - | 5 | 1041 | ||
| Gene Symbol: LOC101929479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017022812.1 | XP_016878301.1 | 5 | 1228 | ||

GOLGA2P10
Genomic Map