This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ACSF2 SIRNA Assays ›
Gene Symbol:
ACSF2
Gene Name:
acyl-CoA synthetase family member 2
Gene Aliases:
ACSMW, AVYV493
Chromosome Location:
Chr.17: 50426158 - 50474845 on Build GRCh38
UniGene ID:
Hs.288959
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 32389

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ACSF2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288968.1 NP_001275897.1 3 483
NM_001288969.1 NP_001275898.1 2 408
NM_001288970.1 NP_001275899.1 2 408
NM_001288972.1 NP_001275901.1 2 408
NM_025149.5 NP_079425.3 2 408
XM_006722110.3 XP_006722173.1 2 348
XM_011525294.1 XP_011523596.1 2 348
GenBank mRNA AK024573.1 - 2 315
AK225044.1 - 2 315
AK225561.1 - 2 314
AK294205.1 - 3 425
AK295258.1 - 2 316
AK300145.1 - 2 316
AY358660.1 - 2 344
BC012053.1 - 2 321 4577089
BC014123.2 - 2 328 4560029
DC296100.1 - 2 348

Back To Top

More Information


Panther Classification:

Molecular Function -

ligase

Gene Ontology Categories:

Function(s) Process(es)

fatty acid metabolic process
long-chain fatty-acyl-CoA biosynthetic process
acyl-CoA ligase activity
protein binding
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-b7w9w:80/100.66.77.211:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline