This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PAAF1 SIRNA Assays ›
Gene Symbol:
PAAF1
Gene Name:
proteasomal ATPase associated factor 1
Gene Aliases:
PAAF, Rpn14, WDR71
Chromosome Location:
Chr.11: 73876699 - 73927736 on Build GRCh38
UniGene ID:
Hs.525017
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 32394

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PAAF1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267806.1 NP_001254735.1 2, 3 92
NM_025155.2 NP_079431.1 2, 3 149
XM_005274309.1 XP_005274366.1 1, 2 49
GenBank mRNA AK021910.1 - 2, 3 130
AK222501.1 - 2, 3 130
AK225520.1 - 2, 3 117
AK298258.1 - 2, 3 92
BC006142.2 - 2, 3 102 3937356
BC021541.1 - 2, 3 113 3942680
DC354686.1 - 2, 3 151

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

viral process
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-98cc9b549-qbh9s:80/100.66.78.193:80.
git-commit: 5e7f4b920385fdbbb5f8d978f1c7c2eb3f871195
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.1-Offline