This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CCDC93 SIRNA Assays ›
Gene Symbol:
CCDC93
Gene Name:
coiled-coil domain containing 93
Gene Aliases:
-
Chromosome Location:
Chr.2: 117915478 - 118014163 on Build GRCh38
UniGene ID:
Hs.107845
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 33225

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CCDC93
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019044.4 NP_061917.3 24 5792
XM_011511359.1 XP_011509661.1 24 5789
XM_011511361.1 XP_011509663.1 23 5546
XM_011511362.1 XP_011509664.1 23 5660
GenBank mRNA AB209993.1 - 24 5710
AK124683.1 - 1 1836
AY726577.1 - 1 1756
BC005078.1 - 1 280
BC080559.1 - 1 262

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Gene Ontology Categories:

Function(s) Process(es)

Golgi to plasma membrane transport
protein transport
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-kbrl8:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline