siRNA Details
| Gene Symbol: THAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031435.3 | NP_113623.1 | 2 | 760 | ||
| GenBank mRNA | AK313452.1 | - | 2 | 322 | ||
| AL136607.1 | - | 2 | 657 | |||
| BC008358.1 | - | 2 | 500 | 3606376 | ||
| CR533485.1 | - | 2 | 147 | |||

THAP2
Genomic Map