This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MFSD14B SIRNA Assays ›
Gene Symbol:
MFSD14B
Gene Name:
major facilitator superfamily domain containing 14B
Gene Aliases:
HIATL1
Chromosome Location:
Chr.9: 94374327 - 94460920 on Build GRCh38
UniGene ID:
Hs.555996
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 34492

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MFSD14B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032558.2 NP_115947.2 12 2210
XM_006717305.3 XP_006717368.1 11 2087
XM_011519114.2 XP_011517416.1 12 1901
XM_017015217.1 XP_016870706.1 12 2011
XM_017015218.1 XP_016870707.1 12 1938
XM_017015219.1 XP_016870708.1 11 1748
XM_017015220.1 XP_016870709.1 11 1762
XM_017015221.1 XP_016870710.1 11 1803
GenBank mRNA AF255650.1 - 6 1184
AK027659.1 - 8 1514
AL832415.1 - 5 1077
BC106062.1 - 12 1947 6162855
BX648846.1 - 9 1621

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transmembrane transport
transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5p4xw:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline