siRNA Details
| Gene Symbol: MAEL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286378.1 | NP_001273307.1 | 3 | 189 | ||
| NM_032858.2 | NP_116247.1 | 2 | 416 | |||
| XM_006711583.1 | XP_006711646.1 | 3 | 220 | |||
| XM_011510067.1 | XP_011508369.1 | 2 | 391 | |||
| XM_011510068.1 | XP_011508370.1 | 3 | 182 | |||
| XM_017002602.1 | XP_016858091.1 | 4 | 557 | |||
| XM_017002603.1 | XP_016858092.1 | 3 | 219 | |||
| GenBank mRNA | AK027810.1 | - | 2 | 223 | ||
| AK131066.1 | - | 2 | 226 | |||
| BC028595.2 | - | 2 | 196 | 5270252 | ||
| DQ076156.2 | - | 2 | 225 | |||
| GU383114.1 | - | 3 | 189 | |||

MAEL
Genomic Map