siRNA Details
| Gene Symbol: SLC18B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052831.2 | NP_439896.1 | 2 | 259 | ||
| XM_017010236.1 | XP_016865725.1 | 2 | 450 | |||
| GenBank mRNA | AK124442.1 | - | 2 | 138 | ||
| AK289918.1 | - | 2 | 278 | |||
| AK293853.1 | - | 2 | 213 | |||
| BC069570.1 | - | 2 | 179 | 7262298 | ||
| CD635228.1 | - | 2 | 259 | |||

SLC18B1
Genomic Map