This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SEPT1 SIRNA Assays ›
Gene Symbol:
SEPT1
Gene Name:
septin 1
Gene Aliases:
DIFF6, LARP, PNUTL3, SEP1
Chromosome Location:
Chr.16: 30378133 - 30382850 on Build GRCh38
UniGene ID:
Hs.632176
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 35399

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SEPT1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052838.4 NP_443070.5 6 528
XM_017022991.1 XP_016878480.1 6 508
GenBank mRNA AF085235.1 - 5 362
AK301045.1 - 5 360
AK303390.1 - 6 491
AL833004.1 - 2 107
AY034176.1 - 5 362
BC012161.1 - 4 350 4576220
BQ072902.1 - 6 541

Back To Top

More Information


Panther Classification:

Molecular Function -

cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
cell division
protein binding
GTP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-xg9l8:80/100.66.77.189:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline