This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DMRT2 SIRNA Assays ›
Gene Symbol:
DMRT2
Gene Name:
doublesex and mab-3 related transcription factor 2
Gene Aliases:
-
Chromosome Location:
Chr.9: 1003237 - 1057554 on Build GRCh38
UniGene ID:
Hs.59506
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 3577

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DMRT2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130865.2 NP_001124337.1 6 1760
NM_006557.6 NP_006548.1 4 2321
NM_181872.4 NP_870987.2 4 1553
XM_011517687.1 XP_011515989.1 6 4101
XM_011517690.2 XP_011515992.1 5 2181
XM_011517694.2 XP_011515996.1 3 1712
XM_017014213.1 XP_016869702.1 6 2503
XM_017014214.1 XP_016869703.1 5 2178
XM_017014215.1 XP_016869704.1 4 2022
GenBank mRNA AF284223.1 - 3 1848
AF284224.1 - 4 1431
AF284225.1 - 3 1353
AK290794.1 - 6 2066
BC030204.1 - 6 1760
BC136493.1 - 3 1432 9020482
BX647565.1 - 5 1367

Back To Top

More Information


Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
sex differentiation
biological_process
regulation of somitogenesis
positive regulation of transcription from RNA polymerase II promoter
embryonic skeletal system development
positive regulation of myotome development
RNA polymerase II regulatory region sequence-specific DNA binding
core promoter proximal region sequence-specific DNA binding
molecular_function
transcription factor activity, sequence-specific DNA binding
protein homodimerization activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-v4fgr:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline